Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100018494.mm.1 |
NR_045151.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018494.mm.1 |
NR_045151.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018494.mm.1 |
TC0100002303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018496.mm.1 |
NR_045151.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018496.mm.1 |
NR_045151.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018496.mm.1 |
TC0100002303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018497.mm.1 |
NR_045151.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018497.mm.1 |
NR_045151.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018497.mm.1 |
TC0100002303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018495.mm.1 |
NR_045151.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018495.mm.1 |
NR_045151.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018495.mm.1 |
TC0100002303.mm.1 |
MTA TC |