Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NM_001252476.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NM_001252476.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NM_001252477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NM_001252477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NM_019830.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NM_019830.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NR_045521.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
NR_045521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026721.mm.1 |
TC0700002980.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026722.mm.1 |
NR_045521.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026722.mm.1 |
NR_045521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026722.mm.1 |
TC0700002980.mm.1 |
MTA TC |