Assay Details
| Gene Symbol: Ccdc77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045558.1 | 1 - 2 | 284 | 98 | ||
| GenBank mRNA | AK031590.1 | - | 1 - 2 | 285 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysine (K)-specific demethylase 5A | Kdm5a |
AA409370, C76986, Jarid1a, RBP2, Rbbp2 |
Anti-sense | Chr. 6 - 120364099 - 120444574 on NCBI Build GRCm38 | |
| Downstream | interleukin 17 receptor A | Il17ra |
AW538159, Cdw217, Il17r, VDw217 |
98.8kb | Anti-sense | Chr. 6 - 120463181 - 120483731 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc77
Genomic Map