Assay Details
| Gene Symbol: Tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045606.1 | 4 - 5 | 648 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB3D, member RAS oncogene family |
Rab3d | C130057E11Rik | -2.8kb | Sense | Chr. 9 - 21907491 - 21918197 on NCBI Build GRCm38 |
| Downstream | coiled-coil domain containing 159 | Ccdc159 | 2510048L02Rik | 4.2kb | Anti-sense | Chr. 9 - 21927471 - 21936256 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tmem205
Genomic Map