Assay Details
| Gene Symbol: 1700028E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045700.1 | 3 - 4 | 511 | 79 | ||
| GenBank mRNA | AK006454.1 | - | 3 - 4 | 468 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | StAR-related lipid transfer (START) domain containing 13 | Stard13 |
DLC2, GT650 |
Anti-sense | Chr. 5 - 151037515 - 151428256 on NCBI Build GRCm38 | |
| Downstream | predicted gene 3704 | Gm3704 | 9.9kb | Sense | Chr. 5 - 151423987 - 151432127 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700028E10Rik
Genomic Map