Assay Details
| Gene Symbol: Gm9833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045710.1 | 1 | 1290 | 126 | ||
| GenBank mRNA | BC060946.1 | - | 1 | 1290 | 6741255 | 126 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
fatty acid binding protein 5, epidermal |
Fabp5 |
E-FABP, Fabpe, Klbp, PA-FABP, mal1 |
-71.7kb | Sense | Chr. 3 - 10012585 - 10016610 on NCBI Build GRCm38 |
| Downstream | peripheral myelin protein 2 | Pmp2 | P2 | 90.2kb | Anti-sense | Chr. 3 - 10179851 - 10183885 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm9833
Genomic Map