Assay Details
| Gene Symbol: Gm10872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045747.1 | 1 - 2 | 52 | 62 | ||
| GenBank mRNA | AK155479.1 | - | 1 - 2 | 53 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding) |
Grina |
1110025J15Rik, Lag, Tmbim3 |
-18.3kb | Anti-sense | Chr. 15 - 76246740 - 76249911 on NCBI Build GRCm38 |
| Downstream | microRNA 3079 | Mir3079 | mmu-mir-3079 | 21.0kb | Sense | Chr. 15 - 76290879 - 76290968 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155479.1 transcripts.
Set Membership: |
|

Gm10872
Genomic Map