Assay Details
Gene Symbol: Gm10787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045882.1 | 2 - 3 | 570 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051077 | AK143813.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121811 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051077 | AK143813.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121811 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08926736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08359937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08359939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08359630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08783572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08382439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08382434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08359941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08840771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08648215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08926738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08638342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08638346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265390 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08638358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08909627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27265405 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1000005416.mm.1 | NR_045882.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005416.mm.1 | NR_045882.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005416.mm.1 | TC1000000751.mm.1 | MTA TC |
