Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400005439.mm.1 |
NR_046041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005439.mm.1 |
NR_046041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005439.mm.1 |
NR_046042.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005439.mm.1 |
NR_046042.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005439.mm.1 |
TC0400000772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400005440.mm.1 |
NR_046041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005440.mm.1 |
NR_046041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005440.mm.1 |
TC0400000772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400005442.mm.1 |
NR_046041.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005442.mm.1 |
NR_046041.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005442.mm.1 |
NR_046042.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005442.mm.1 |
NR_046042.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005442.mm.1 |
TC0400000772.mm.1 |
MTA TC |