Assay Details
| Gene Symbol: 7420700N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046272.1 | 2 - 3 | 265 | 82 | ||
| GenBank mRNA | AK018363.1 | - | 2 - 3 | 265 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fucosyltransferase 10 | Fut10 |
fuc-TX, fut10A |
-116.9kb | Sense | Chr. 8 - 31187331 - 31261924 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 38572 |
Gm38572 | 219.1kb | Anti-sense | Chr. 8 - 31598979 - 31663951 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1600001809.rn
Set Membership: |
|

7420700N18Rik
Genomic Map