Assay Details
Gene Symbol: Prl8a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073166.1 | 1 - 2 | 134 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
prolactin family 2, subfamily b, member 1 |
Prl2b1 |
2310047B08Rik, PLP-K, Plpk, Prlpk, mPLP-K |
-46.2kb | Sense | Chr. 13 - 27383345 - 27390846 on NCBI Build GRCm38 |
Downstream |
prolactin family 8, subfamily a, member 81 |
Prl8a8 |
1600032B14Rik, AV036230, PLP-Cgamma, Prlpc3 |
68.3kb | Sense | Chr. 13 - 27507032 - 27513228 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
