Assay Details
| Gene Symbol: Gabpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073183.1 | 3 - 4 | 508 | 91 | ||
| GenBank mRNA | AY282804.1 | - | 3 - 4 | 354 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | histidine decarboxylase | Hdc |
AW108189, Hdc-a, Hdc-c, Hdc-e, Hdc-s |
-26.0kb | Sense | Chr. 2 - 126593659 - 126624022 on NCBI Build GRCm38 |
| Downstream | ubiquitin specific peptidase 8 | Usp8 |
AI574262, AW557536, Ubpy, mKIAA0055 |
53.5kb | Anti-sense | Chr. 2 - 126707202 - 126759314 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gabpb1
Genomic Map