Assay Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073186.1 | 2 - 3 | 504 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102636700 | LOC102636700 | -10.6kb | Anti-sense | Chr. 9 - 75310333 - 75314383 on NCBI Build GRCm38 | |
| Downstream | Bcl2-like 10 | Bcl2l10 |
AA420380, AU023065, Boo, C85687, Diva |
20.5kb | Sense | Chr. 9 - 75347758 - 75351640 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gnb5
Genomic Map