Assay Details
Gene Symbol: Rpph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002142.2 | 1 | 268 | 164 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin B1 interacting protein 1 | Ccnb1ip1 |
Gm288, Hei10, mei4 |
-11.5kb | Sense | Chr. 14 - 50789249 - 50795983 on NCBI Build GRCm38 |
Downstream |
poly (ADP-ribose) polymerase family, member 2 |
Parp2 |
ARTD2, Adprt2, Adprtl2, Aspartl2, C78626, PARP-2 |
127.0 | Anti-sense | Chr. 14 - 50807898 - 50821301 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC1400001987.mm
This assay targets the same location as RTA transcript cluster(s) TC1400001601.rn
This assay targets the same location as RTA transcript cluster(s) TC1400001601.rn
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
