Assay Details
| Gene Symbol: Snora52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034049.2 | 1 | 66 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | p53 induced death domain protein 1 | Pidd1 |
1200011D09Rik, AU042446, Lrdd, Pidd |
-4.4kb | Anti-sense | Chr. 7 - 141438113 - 141444408 on NCBI Build GRCm38 |
| Downstream | patatin-like phospholipase domain containing 2 | Pnpla2 |
0610039C21Rik, 1110001C14Rik, Atgl, TTS-2.2 |
6.3kb | Sense | Chr. 7 - 141455188 - 141460743 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snora52
Genomic Map