Assay Details
| Gene Symbol: Gm2837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040388.1 | 1 | 640 | 69 | ||
| GenBank mRNA | BC117727.1 | - | 1 | 640 | 40089638 | 69 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 2825 | Gm2825 |
GMCL1P1, Gclh, Gmcl1l |
-80.4kb | Sense | Chr. X - 32973897 - 32975846 on NCBI Build GRCm38 |
| Downstream | predicted gene 2863 | Gm2863 | 256.3kb | Sense | Chr. X - 33313338 - 33315288 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm2837
Genomic Map