Assay Details
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253738.1 | NP_001240667.1 | 6 - 7 | 805 | 117 | |
NM_001271764.1 | NP_001258693.1 | 6 - 7 | 805 | 117 | ||
NM_022314.3 | NP_071709.3 | 7 - 8 | 788 | 117 | ||
GenBank mRNA | AK050665.1 | - | 6 - 7 | 651 | 117 | |
AK088111.1 | - | 6 - 7 | 665 | 117 | ||
AK088902.1 | - | 6 - 7 | 663 | 117 | ||
AK145238.1 | - | 6 - 7 | 662 | 117 | ||
AK145243.1 | - | 6 - 7 | 662 | 117 | ||
AK145804.1 | - | 6 - 7 | 662 | 117 | ||
AK146349.1 | - | 6 - 7 | 635 | 117 | ||
AK146557.1 | - | 6 - 7 | 787 | 117 | ||
AK151441.1 | - | 6 - 7 | 687 | 117 | ||
AK151500.1 | - | 6 - 7 | 662 | 117 | ||
AK151571.1 | - | 6 - 7 | 661 | 117 | ||
AK151578.1 | - | 6 - 7 | 662 | 117 | ||
AK151595.1 | - | 6 - 7 | 634 | 117 | ||
AK151602.1 | - | 6 - 7 | 662 | 117 | ||
AK151991.1 | - | 6 - 7 | 661 | 117 | ||
AK152260.1 | - | 6 - 7 | 661 | 117 | ||
AK152659.1 | - | 6 - 7 | 634 | 117 | ||
AK153031.1 | - | 6 - 7 | 634 | 117 | ||
AK160011.1 | - | 6 - 7 | 687 | 117 | ||
AK165428.1 | - | 6 - 7 | 661 | 117 | ||
AK166836.1 | - | 6 - 7 | 649 | 117 | ||
AK167268.1 | - | 6 - 7 | 639 | 117 | ||
AK167534.1 | - | 6 - 7 | 667 | 117 | ||
AK167581.1 | - | 6 - 7 | 662 | 117 | ||
AK168843.1 | - | 6 - 7 | 630 | 117 | ||
AK169102.1 | - | 6 - 7 | 633 | 117 | ||
AK169245.1 | - | 6 - 7 | 638 | 117 | ||
AK169821.1 | - | 6 - 7 | 679 | 117 | ||
BC029186.1 | - | 7 - 8 | 745 | 4502880 | 117 | |
BC055407.1 | - | 3931 | 117 | |||
BC092045.1 | - | 6 - 7 | 638 | 3591372 | 117 | |
X53753.1 | - | 6 - 7 | 673 | 117 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
