Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
NM_001253781.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
NM_001253781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
NM_001253782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
NM_001253782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
NM_011169.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
NM_011169.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000700.mm.1 |
TC1500002327.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
NM_001253781.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
NM_001253781.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
NM_001253782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
NM_001253782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
NM_011169.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
NM_011169.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500000702.mm.1 |
TC1500002327.mm.1 |
MTA TC |