Assay Details
| Gene Symbol: Marcks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008538.2 | NP_032564.1 | 1 - 2 | 493 | 61 | |
| GenBank mRNA | BC002068.1 | - | 1 - 2 | 506 | 61 | |
| BC039911.1 | - | 1 - 2 | 498 | 61 | ||
| BC046601.1 | - | 1 - 2 | 454 | 6311439 | 61 | |
| M60474.1 | - | 1 - 2 | 482 | 61 | ||
| X59374.1 | - | 1 - 2 | 384 | 61 | ||
More Information
Set Membership: |
|

Marcks
Genomic Map