Assay Details
| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025093.2 | NP_001020264.1 | 15 | 3975 | 112 | |
| NM_001284369.1 | NP_001271298.1 | 13 | 3815 | 112 | ||
| NM_001284372.1 | NP_001271301.1 | 13 | 3681 | 112 | ||
| NM_001284373.1 | NP_001271302.1 | 12 | 3618 | 112 | ||
| NM_001284374.1 | NP_001271303.1 | 12 | 3700 | 112 | ||
| NM_001284376.1 | NP_001271305.1 | 15 | 3857 | 112 | ||
| NM_009715.3 | NP_033845.1 | 14 | 3878 | 112 | ||
| GenBank mRNA | AK144977.1 | - | 13 | 3633 | 112 | |
| AK145887.1 | - | 14 | 3257 | 112 | ||
| BC042210.1 | - | 12 | 3630 | 4018323 | 112 | |
| BC079883.1 | - | 14 | 3800 | 6839572 | 112 | |
| BC082596.1 | - | 12 | 3565 | 30649207 | 112 | |
| BC085137.1 | - | 12 | 3642 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atf2
Genomic Map