Assay Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301340.1 | NP_001288269.1 | 16 | 2245 | 102 | |
| NM_001301341.1 | NP_001288270.1 | 18 | 2372 | 102 | ||
| NM_001301343.1 | NP_001288272.1 | 16 | 2231 | 102 | ||
| NM_001301344.1 | NP_001288273.1 | 15 | 2173 | 102 | ||
| NM_001301345.1 | NP_001288274.1 | 15 | 2113 | 102 | ||
| NM_025279.3 | NP_079555.1 | 16 | 2185 | 102 | ||
| GenBank mRNA | AK088462.1 | - | 15 | 2099 | 102 | |
| AK128891.1 | - | 16 | 2182 | 102 | ||
| AK135519.1 | - | 16 | 2158 | 102 | ||
| AK150848.1 | - | 16 | 2225 | 102 | ||
| AK151689.1 | - | 15 | 2126 | 102 | ||
| AK152927.1 | - | 15 | 2127 | 102 | ||
| AK160887.1 | - | 16 | 2186 | 102 | ||
| AK166656.1 | - | 16 | 2249 | 102 | ||
| AK166927.1 | - | 16 | 2203 | 102 | ||
| AK167600.1 | - | 16 | 2175 | 102 | ||
| AK168918.1 | - | 12 | 2086 | 102 | ||
| AK169064.1 | - | 16 | 2234 | 102 | ||
| BC089328.1 | - | 16 | 2169 | 6402614 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpk
Genomic Map