| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300013364.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013363.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013363.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013367.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013367.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013372.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013372.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013373.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013373.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013358.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013361.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013361.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013404.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013366.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013366.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013362.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013362.mm.1 |
TC1300002099.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013365.mm.1 |
NM_025279.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013365.mm.1 |
TC1300002099.mm.1 |
MTA TC |