Assay Details
| Gene Symbol: Smim10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073368.1 | 2 - 3 | 538 | 77 | ||
| GenBank mRNA | AA048533.1 | - | 2 - 3 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
taste receptor, type 2, member 140 |
Tas2r140 |
T2R40, T2r34, TRB3, TRB5, Tas2r13, Tas2r137, Tas2r40, Tas2r8, mTRB3, mt2r64 |
-50.6kb | Anti-sense | Chr. 6 - 133054855 - 133055793 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 5530400C23 gene | 5530400C23Rik | GRP | 184.2kb | Sense | Chr. 6 - 133292216 - 133295790 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Smim10l1
Genomic Map