Assay Details
Gene Symbol: Gm38404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073373.1 | 3 - 4 | 566 | 71 | ||
GenBank mRNA | AK051577.1 | - | 3 - 4 | 567 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 9330179D12 gene | 9330179D12Rik | -83.8kb | Anti-sense | Chr. 6 - 127149389 - 127212419 on NCBI Build GRCm38 | |
Downstream |
poly (ADP-ribose) polymerase family, member 11 |
Parp11 |
5330431N24Rik, AI851877, ARTD11, HIN1L |
148.5kb | Anti-sense | Chr. 6 - 127446845 - 127512908 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
