Assay Details
Gene Symbol: 4930577N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073429.1 | 1 | 122 | 80 | ||
GenBank mRNA | AK086057.1 | - | 1 | 64 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CCR4 carbon catabolite repression 4-like (S. cerevisiae) | Ccrn4l |
AU043840, Ccr4, nocturnin |
-25.1kb | Sense | Chr. 3 - 51224447 - 51251654 on NCBI Build GRCm38 |
Downstream | predicted gene 5103 | Gm5103 | A630062H14 | 99.6kb | Sense | Chr. 3 - 51378025 - 51386324 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
