Assay Details
| Gene Symbol: Gpr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_074084.1 | 1 - 2 | 153 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cAMP responsive element binding protein-like 2 | Crebl2 |
AI046348, B230205M03 |
-28.0kb | Anti-sense | Chr. 6 - 134828686 - 134858931 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2810454H06 gene | 2810454H06Rik | 10.2kb | Anti-sense | Chr. 6 - 134897961 - 134900785 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr19
Genomic Map