Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300003453.mm.1 |
NM_199056.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003453.mm.1 |
NR_075085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003453.mm.1 |
TC1300000563.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300003458.mm.1 |
NR_075085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003458.mm.1 |
TC1300000563.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300003456.mm.1 |
NM_199056.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003456.mm.1 |
NR_075085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003456.mm.1 |
TC1300000563.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300003454.mm.1 |
NM_199056.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003454.mm.1 |
NR_075085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003454.mm.1 |
TC1300000563.mm.1 |
MTA TC |