Assay Details
Gene Symbol: C430002N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102294.1 | 1 - 2 | 153 | 70 | ||
GenBank mRNA | DV646733.1 | - | 1 - 2 | 155 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA E030011O05 gene | E030011O05Rik | -12.7kb | Anti-sense | Chr. 9 - 96731575 - 96752831 on NCBI Build GRCm38 | |
Downstream | 2-phosphoxylose phosphatase 1 | Pxylp1 |
9430094M07Rik, Acpl2, BB177120, C130099A20Rik |
53.2kb | Anti-sense | Chr. 9 - 96823336 - 96892680 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
