Assay Details
Gene Symbol: D630024D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102312.1 | 3 - 4 | 523 | 125 | ||
NR_102314.1 | 2 - 3 | 371 | 125 | |||
GenBank mRNA | AK085420.1 | - | 3 - 4 | 501 | 125 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | biorientation of chromosomes in cell division 1 | Bod1 |
2310022M17Rik, BOD2.1, Fam44b |
-129.1kb | Sense | Chr. 11 - 31665150 - 31671862 on NCBI Build GRCm38 |
Downstream | cytoplasmic polyadenylation element binding protein 4 | Cpeb4 |
Cpe-bp4, Cpeb-4, mCPEB-4 |
68.4kb | Anti-sense | Chr. 11 - 31870884 - 31935635 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
