Assay Details
Gene Symbol: 6430710C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102348.1 | 1 - 2 | 150 | 84 | ||
NR_102349.1 | 1 - 2 | 150 | 84 | |||
GenBank mRNA | AK018302.1 | - | 1 - 2 | 106 | 84 | |
AK019889.1 | - | 1 - 2 | 150 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 8430437L04 gene | 8430437L04Rik | -70.6kb | Anti-sense | Chr. 2 - 72703510 - 72729468 on NCBI Build GRCm38 | |
Downstream | trans-acting transcription factor 3 | Sp3 | D130027J01Rik | 122.7kb | Sense | Chr. 2 - 72936430 - 72980446 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
