Assay Details
Gene Symbol: Zbtb24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102360.1 | 3 - 4 | 1321 | 88 | ||
GenBank mRNA | AK154467.1 | - | 3 - 4 | 1297 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adenylate kinase 9 | Ak9 |
Akd1, Akd2, Gm234, Gm7127 |
-21.5kb | Sense | Chr. 10 - 41303420 - 41433568 on NCBI Build GRCm38 |
Downstream |
microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Mical1 |
MICAL, MICAL-1, Nical |
20.1kb | Sense | Chr. 10 - 41475860 - 41487030 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
