Assay Details
| Gene Symbol: Dmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102372.1 | 1 - 2 | 101 | 80 | ||
| GenBank mRNA | HM802239.1 | - | 1 - 2 | 101 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | brain protein I3 | Bri3 | I3 | Sense | Chr. 5 - 144244437 - 144446757 on NCBI Build GRCm38 | |
| Downstream | neuronal pentraxin 2 | Nptx2 |
Narp, np2 |
184.9kb | Sense | Chr. 5 - 144545887 - 144557478 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Dmr
Genomic Map