Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102722.1 | 4 - 5 | 397 | 107 | ||
| GenBank mRNA | AK041230.1 | - | 4 - 5 | 332 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | free fatty acid receptor 1 | Ffar1 | Gpr40 | -16.0kb | Sense | Chr. 7 - 30860568 - 30861470 on NCBI Build GRCm38 |
| Downstream |
DNA segment, Chr 7, ERATO Doi 128, expressed |
D7Ertd128e | 3.8kb | Anti-sense | Chr. 7 - 30881953 - 30886420 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Cd22
Genomic Map