Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102723.1 | 7 - 8 | 1446 | 94 | ||
| GenBank mRNA | BC046957.1 | - | 7 - 8 | 1358 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | free fatty acid receptor 1 | Ffar1 | Gpr40 | -11.3kb | Sense | Chr. 7 - 30860568 - 30861470 on NCBI Build GRCm38 |
| Downstream |
DNA segment, Chr 7, ERATO Doi 128, expressed |
D7Ertd128e | 8.5kb | Anti-sense | Chr. 7 - 30881953 - 30886420 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Cd22
Genomic Map