Assay Details
| Gene Symbol: Dph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104304.1 | 1 - 2 | 279 | 99 | ||
| GenBank mRNA | AK017764.1 | - | 1 - 2 | 197 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15 | Galnt15 |
4631401E18Rik, Galntl2, mpp-GalNAc-T15 |
-22.7kb | Anti-sense | Chr. 14 - 32028651 - 32062204 on NCBI Build GRCm38 |
| Downstream | oxidoreductase NAD-binding domain containing 1 | Oxnad1 |
2410002F01Rik, AW492292, BC019806 |
36.0 | Anti-sense | Chr. 14 - 32085728 - 32103203 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Dph3
Genomic Map