Assay Details
| Gene Symbol: 1700024P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104376.1 | 15 - 16 | 2016 | 120 | ||
| GenBank mRNA | BC144858.1 | - | 15 - 16 | 2017 | 9053406 | 120 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
complement component 8, alpha polypeptide |
C8a | -127.4kb | Anti-sense | Chr. 4 - 104815679 - 104876486 on NCBI Build GRCm38 | |
| Downstream |
protein kinase, AMP-activated, alpha 2 catalytic subunit |
Prkaa2 |
2310008I11Rik, A830082D05, AMPKalpha2 |
24.3kb | Anti-sense | Chr. 4 - 105029650 - 105109898 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700024P16Rik
Genomic Map