Assay Details
| Gene Symbol: Clmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104435.1 | 11 - 12 | 2964 | 68 | ||
| NR_104436.1 | 11 - 12 | 2964 | 68 | |||
| NR_104437.1 | 11 - 12 | 2964 | 68 | |||
| NR_104438.1 | 11 - 12 | 2964 | 68 | |||
| GenBank mRNA | AB059645.1 | - | 11 - 12 | 2957 | 68 | |
| AB059646.1 | - | 11 - 12 | 2957 | 68 | ||
| AB059647.1 | - | 11 - 12 | 2957 | 68 | ||
| AB059648.1 | - | 11 - 12 | 2957 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
dicer 1, ribonuclease type III |
Dicer1 |
1110006F08Rik, D12Ertd7e, mKIAA0928 |
-22.5kb | Sense | Chr. 12 - 104687740 - 104751952 on NCBI Build GRCm38 |
| Downstream |
spectrin repeat containing, nuclear envelope family member 3 |
Syne3 | nesprin-3 | 153.9kb | Sense | Chr. 12 - 104929830 - 105010098 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Clmn
Genomic Map