Assay Details
| Gene Symbol: Galnt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104449.1 | 4 - 5 | 557 | 70 | ||
| GenBank mRNA | AK170381.1 | - | 4 - 5 | 543 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A630023P12 gene | A630023P12Rik | -91.9kb | Sense | Chr. 5 - 110514923 - 110525980 on NCBI Build GRCm38 | |
| Downstream | nucleolar complex associated 4 homolog (S. cerevisiae) | Noc4l | AI326906 | 29.1kb | Anti-sense | Chr. 5 - 110648419 - 110653382 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt9
Genomic Map