Assay Details
| Gene Symbol: Rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104478.1 | 1 - 2 | 36 | 64 | ||
| GenBank mRNA | CV782943.1 | - | 1 - 2 | 36 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycoprotein 9 (platelet) | Gp9 |
Cd42, GPIX |
-14.8kb | Anti-sense | Chr. 6 - 87776122 - 87779786 on NCBI Build GRCm38 |
| Downstream | ISY1 splicing factor homolog (S. cerevisiae) | Isy1 |
5830446M03Rik, AI181014, AU020769 |
6.3kb | Sense | Chr. 6 - 87818447 - 87838759 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rab43
Genomic Map