Assay Details
| Gene Symbol: Gm29805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105054.1 | 2 - 3 | 437 | 92 | ||
| GenBank mRNA | AK045951.1 | - | 2 - 3 | 438 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm29805
Genomic Map