Assay Details
Gene Symbol: Mir6371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105790.1 | 1 | 54 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cytochrome c, somatic |
Cycs | -5.9kb | Anti-sense | Chr. 6 - 50562563 - 50566474 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4921507P07 gene | 4921507P07Rik | TISP74 | 794.0 | Anti-sense | Chr. 6 - 50573302 - 50596912 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
