Assay Details
| Gene Symbol: Mir6405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105831.1 | 1 | 24 | 121 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26644 |
Gm26644 | -113.0kb | Anti-sense | Chr. 19 - 44685753 - 44695379 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1700039E22 gene | 1700039E22Rik | 20.0kb | Anti-sense | Chr. 19 - 44828493 - 44835938 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mir6405
Genomic Map