Assay Details
| Gene Symbol: Mir6916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105881.1 | 1 | 27 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Riken cDNA A430046D13 gene | A430046D13Rik | Anti-sense | Chr. 10 - 128493980 - 128513755 on NCBI Build GRCm38 | ||
| Downstream | zinc finger CCCH type containing 10 | Zc3h10 | AI790326 | 31.5kb | Sense | Chr. 10 - 128543563 - 128547744 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6916
Genomic Map