Assay Details
| Gene Symbol: Mir6919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105884.1 | 1 | 31 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36556 |
Gm36556 | -7.1kb | Anti-sense | Chr. 11 - 50222140 - 50225172 on NCBI Build GRCm38 | |
| Downstream | leukotriene C4 synthase | Ltc4s | 2.7kb | Anti-sense | Chr. 11 - 50235003 - 50239175 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6919
Genomic Map