Assay Details
| Gene Symbol: Mir6996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105962.1 | 1 | 28 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 0610009E02 gene | 0610009E02Rik | -11.8kb | Anti-sense | Chr. 2 - 26446041 - 26458268 on NCBI Build GRCm38 | |
| Downstream | EGF-like domain 7 | Egfl7 |
VE-statin, Zneu1 |
110.9kb | Anti-sense | Chr. 2 - 26581028 - 26592682 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6996
Genomic Map