Assay Details
| Gene Symbol: Mir7012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105979.1 | 1 | 29 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DENN/MADD domain containing 4B | Dennd4b | Sense | Chr. 3 - 90264920 - 90280665 on NCBI Build GRCm38 | ||
| Downstream | GATA zinc finger domain containing 2B | Gatad2b |
AL118180, C430014D17Rik, P66beta, mKIAA1150 |
23.1kb | Sense | Chr. 3 - 90293285 - 90363406 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7012
Genomic Map