Assay Details
| Gene Symbol: Mir7224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106083.1 | 1 | 36 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1 |
B3galt1 | beta3Gal-T1 | Sense | Chr. 2 - 67565962 - 68122882 on NCBI Build GRCm38 | |
| Downstream | microRNA 6335 | Mir6335 |
Gm27836, mmu-mir-6335 |
105.5kb | Sense | Chr. 2 - 67781043 - 67781140 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7224
Genomic Map