Assay Details
Gene Symbol: Mir7669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106129.1 | 1 | 28 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 19710 |
Gm19710 | -9.8kb | Anti-sense | Chr. 3 - 89998760 - 90001325 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4933434E20 gene | 4933434E20Rik |
5730552F22Rik, AI462154, NICE-3, NS5ATP4 |
40.4kb | Anti-sense | Chr. 3 - 90051636 - 90063341 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
