Assay Details
Gene Symbol: 1700065L07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108032.1 | 2 - 3 | 234 | 82 | ||
GenBank mRNA | AK006896.1 | - | 2 - 3 | 237 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
succinate-CoA ligase, GDP-forming, alpha subunit |
Suclg1 |
1500000I01Rik, Sucla1 |
-163.3kb | Anti-sense | Chr. 6 - 73248505 - 73276907 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4931417E11 gene | 4931417E11Rik | 6.9kb | Sense | Chr. 6 - 73468573 - 73469667 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
