Assay Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110353.1 | 1 - 2 | 95 | 64 | ||
GenBank mRNA | AK030259.1 | - | 1 - 2 | 95 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1 | Riiad1 |
2310007A19Rik, Ncrna00166 |
-7.6kb | Anti-sense | Chr. 3 - 94464984 - 94475078 on NCBI Build GRCm38 |
Downstream | sorting nexin family member 27 | Snx27 |
5730552M22Rik, ESTM45, ESTM47, R75405 |
12.7kb | Anti-sense | Chr. 3 - 94497542 - 94582716 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
